Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
QCR6
Entrez Gene ID
850593
Gene Name
Gene Aliases
COR3, UCR6, YFR033C
Chromosome Location
Chr.VI: 224326 - 224769 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179998.3 NP_116691.3 1 - 1 350 91

View Full Product Details