Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AIM14
Entrez Gene ID
852716
Gene Name
Gene Aliases
YGL160W, YNO1
Chromosome Location
Chr.VII: 200561 - 202273 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181025.1 NP_011355.1 1 - 1 1551 87

View Full Product Details