Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GTS1
Entrez Gene ID
852694
Gene Name
Gene Aliases
FHT1, LSR1, YGL181W
Chromosome Location
Chr.VII: 157906 - 159096 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181046.1 NP_011334.1 1 - 1 842 74

View Full Product Details