Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERV1
Entrez Gene ID
852916
Gene Name
Gene Aliases
YGR029W
Chromosome Location
Chr.VII: 543553 - 544205 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181158.3 NP_011543.4 1 - 2 87 93

View Full Product Details