Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BUD9
Entrez Gene ID
852932
Gene Name
Gene Aliases
YGR041W
Chromosome Location
Chr.VII: 577487 - 579130 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181170.3 NP_011555.3 1 - 1 1556 108

View Full Product Details