Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VOA1
Entrez Gene ID
852998
Gene Name
Gene Aliases
YGR106C
Chromosome Location
Chr.VII: 698989 - 699786 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181235.1 NP_011620.1 1 - 1 646 96

View Full Product Details