Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHB1
Entrez Gene ID
853033
Gene Name
Gene Aliases
YGR132C
Chromosome Location
Chr.VII: 755589 - 756452 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181261.3 NP_011648.3 1 - 1 649 95

View Full Product Details