Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAY1
Entrez Gene ID
853179
Gene Name
Gene Aliases
YGR263C
Chromosome Location
Chr.VII: 1017967 - 1019241 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181392.3 NP_011779.3 1 - 1 1150 70

View Full Product Details