Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRF1
Entrez Gene ID
856433
Gene Name
Gene Aliases
FIL1, KIM4, YHR038W
Chromosome Location
Chr.VIII: 184066 - 184758 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179168.1 NP_011903.1 1 - 1 543 93

View Full Product Details