Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMF2
Entrez Gene ID
856447
Gene Name
Gene Aliases
YHR050W
Chromosome Location
Chr.VIII: 207654 - 209303 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179180.1 NP_011917.1 1 - 1 1555 87

View Full Product Details