Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KSP1
Entrez Gene ID
856482
Gene Name
Gene Aliases
YHR082C
Chromosome Location
Chr.VIII: 268459 - 271548 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179212.1 NP_011950.1 1 - 1 2944 60

View Full Product Details