Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CIA2
Entrez Gene ID
856522
Gene Name
Gene Aliases
YHR122W
Chromosome Location
Chr.VIII: 353625 - 354320 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179252.1 NP_011990.1 1 - 1 549 77

View Full Product Details