Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KEL1
Entrez Gene ID
856563
Gene Name
Gene Aliases
YHR158C
Chromosome Location
Chr.VIII: 413685 - 417179 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179289.1 NP_012028.1 1 - 1 2947 79

View Full Product Details