Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DOT5
Entrez Gene ID
854805
Gene Name
Gene Aliases
YIL010W
Chromosome Location
Chr.IX: 334882 - 335529 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179360.3 NP_012255.3 1 - 1 550 111

View Full Product Details