Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AVT7
Entrez Gene ID
854721
Gene Name
Gene Aliases
YIL088C
Chromosome Location
Chr.IX: 197931 - 199403 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179436.1 NP_012178.1 1 - 1 1247 74

View Full Product Details