Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOH1
Entrez Gene ID
853413
Gene Name
Gene Aliases
OSW4, YJL038C
Chromosome Location
Chr.X: 375115 - 375774 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181472.1 NP_012496.1 1 - 1 343 70

View Full Product Details