Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IRC8
Entrez Gene ID
853396
Gene Name
Gene Aliases
YJL051W
Chromosome Location
Chr.X: 339787 - 342255 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181484.1 NP_012484.1 1 - 1 2356 92

View Full Product Details