Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDC8
Entrez Gene ID
853520
Gene Name
Gene Aliases
YJR057W
Chromosome Location
Chr.X: 544062 - 544712 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181715.1 NP_012591.1 1 - 1 548 83

View Full Product Details