Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARP3
Entrez Gene ID
853528
Gene Name
Gene Aliases
ACT4, YJR065C
Chromosome Location
Chr.X: 557809 - 559158 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181723.3 NP_012599.3 1 - 1 1248 97

View Full Product Details