Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STE18
Entrez Gene ID
853548
Gene Name
Gene Aliases
YJR086W
Chromosome Location
Chr.X: 586068 - 586400 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181743.1 NP_012619.1 1 - 1 46 131

View Full Product Details