Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTM1
Entrez Gene ID
853827
Gene Name
Gene Aliases
YKL039W
Chromosome Location
Chr.XI: 362622 - 364193 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179605.1 NP_012885.2 1 - 1 1450 101

View Full Product Details