Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KKQ8
Entrez Gene ID
853686
Gene Name
Gene Aliases
YKL168C
Chromosome Location
Chr.XI: 131288 - 133462 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179734.1 NP_012753.2 1 - 1 1849 67

View Full Product Details