Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YSR3
Entrez Gene ID
853927
Gene Name
Gene Aliases
LBP2, YKR053C
Chromosome Location
Chr.XI: 534067 - 535281 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179843.3 NP_012979.3 1 - 1 1142 83

View Full Product Details