Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEF1
Entrez Gene ID
850758
Gene Name
Gene Aliases
YLR069C
Chromosome Location
Chr.XII: 271630 - 273915 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181956.1 NP_013170.1 1 - 1 2050 75

View Full Product Details