Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HOG1
Entrez Gene ID
850803
Gene Name
Gene Aliases
SSK3, YLR113W
Chromosome Location
Chr.XII: 371620 - 372927 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182000.1 NP_013214.1 1 - 1 1248 69

View Full Product Details