Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YPS1
Entrez Gene ID
850811
Gene Name
Gene Aliases
YAP3, YLR120C
Chromosome Location
Chr.XII: 386511 - 388220 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182007.1 NP_013221.1 1 - 1 1547 96

View Full Product Details