Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMX1
Entrez Gene ID
850902
Gene Name
Gene Aliases
YLR205C
Chromosome Location
Chr.XII: 552724 - 553677 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182092.1 NP_013306.2 1 - 1 849 107

View Full Product Details