Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPN13
Entrez Gene ID
851140
Gene Name
Gene Aliases
YLR421C
Chromosome Location
Chr.XII: 965090 - 965560 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182309.3 NP_013525.3 1 - 1 257 101

View Full Product Details