Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
URA5
Entrez Gene ID
854865
Gene Name
Gene Aliases
PYR5, YML106W
Chromosome Location
Chr.XIII: 56773 - 57453 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182468.1 NP_013601.1 1 - 1 455 94

View Full Product Details