Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DFG5
Entrez Gene ID
855278
Gene Name
Gene Aliases
YMR238W
Chromosome Location
Chr.XIII: 746353 - 747729 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182745.1 NP_013965.1 1 - 1 1248 103

View Full Product Details