Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DSK2
Entrez Gene ID
855319
Gene Name
Gene Aliases
YMR276W
Chromosome Location
Chr.XIII: 818827 - 819948 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182783.1 NP_014003.1 1 - 1 1048 63

View Full Product Details