Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSE1
Entrez Gene ID
855356
Gene Name
Gene Aliases
KAP121, YMR308C
Chromosome Location
Chr.XIII: 888952 - 892221 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182819.1 NP_014039.1 1 - 1 3149 101

View Full Product Details