Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BOP3
Entrez Gene ID
855686
Gene Name
Gene Aliases
YNL042W
Chromosome Location
Chr.XIV: 548100 - 549290 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182881.1 NP_014356.1 1 - 1 1049 90

View Full Product Details