Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OCA1
Entrez Gene ID
855624
Gene Name
Gene Aliases
YNL099C
Chromosome Location
Chr.XIV: 438567 - 439283 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182937.3 NP_014300.3 1 - 1 654 100

View Full Product Details