Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HST1
Entrez Gene ID
854086
Gene Name
Gene Aliases
YOL068C
Chromosome Location
Chr.XV: 200368 - 201879 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183323.1 NP_014573.1 1 - 1 1351 62

View Full Product Details