Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THP1
Entrez Gene ID
854082
Gene Name
Gene Aliases
BUD29, YOL072W
Chromosome Location
Chr.XV: 194970 - 196337 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183327.1 NP_014569.1 1 - 1 1247 106

View Full Product Details