Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RTS2
Entrez Gene ID
854244
Gene Name
Gene Aliases
YOR077W
Chromosome Location
Chr.XV: 471899 - 472597 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183496.1 NP_014720.1 1 - 1 552 90

View Full Product Details