Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCS2
Entrez Gene ID
854344
Gene Name
Gene Aliases
YOR173W
Chromosome Location
Chr.XV: 657264 - 658325 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183592.1 NP_014816.2 1 - 1 849 71

View Full Product Details