Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRO2
Entrez Gene ID
854501
Gene Name
Gene Aliases
YOR323C
Chromosome Location
Chr.XV: 921535 - 922905 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183743.1 NP_014968.1 1 - 1 1244 101

View Full Product Details