Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ISM1
Entrez Gene ID
856067
Gene Name
Gene Aliases
YPL040C
Chromosome Location
Chr.XVI: 475993 - 479001 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183854.1 NP_015285.1 1 - 1 2949 92

View Full Product Details