Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSD1
Entrez Gene ID
856000
Gene Name
Gene Aliases
LPG5, YPL104W
Chromosome Location
Chr.XVI: 355700 - 357676 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183918.1 NP_015221.1 1 - 1 1843 90

View Full Product Details