Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KES1
Entrez Gene ID
855958
Gene Name
Gene Aliases
BSR3, LPI3, OSH4, YPL145C
Chromosome Location
Chr.XVI: 278395 - 279699 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183959.1 NP_015180.1 1 - 1 1252 74

View Full Product Details