Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RTC6
Entrez Gene ID
855918
Gene Name
Gene Aliases
TAE4, YPL183W-A, bL36m
Chromosome Location
Chr.XVI: 199095 - 199376 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184317.1 NP_015141.1 1 - 1 149 101

View Full Product Details