Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SSO1
Entrez Gene ID
855844
Gene Name
Gene Aliases
YPL232W
Chromosome Location
Chr.XVI: 107275 - 108147 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184046.1 NP_015092.1 1 - 1 455 72

View Full Product Details