Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KEL3
Entrez Gene ID
855865
Gene Name
Gene Aliases
YPL263C
Chromosome Location
Chr.XVI: 44551 - 46506 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184077.1 NP_015060.1 1 - 1 1849 93

View Full Product Details