Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGC1
Entrez Gene ID
856132
Gene Name
Gene Aliases
YPR021C
Chromosome Location
Chr.XVI: 600649 - 603357 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184118.1 NP_015346.1 1 - 1 2647 98

View Full Product Details