Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VMA13
Entrez Gene ID
856148
Gene Name
Gene Aliases
CLS11, YPR036W
Chromosome Location
Chr.XVI: 643836 - 645272 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184133.1 NP_015361.1 1 - 1 1344 96

View Full Product Details