Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRS2
Entrez Gene ID
856196
Gene Name
Gene Aliases
YPR081C
Chromosome Location
Chr.XVI: 702114 - 703970 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184178.1 NP_015406.1 1 - 1 1744 80

View Full Product Details