Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS0B
Entrez Gene ID
850737
Gene Name
Gene Aliases
NAB1B, YLR048W, YST2
Chromosome Location
Chr.XII: 242232 - 243349 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181935.1 NP_013149.1 2 - 2 649 105

View Full Product Details