Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSTM1
Entrez Gene ID
100049681
Gene Name
osteoclastogenesis associated transmembrane protein 1
Gene Aliases
GL
Chromosome Location
Chr.1: 74139326 - 74181165 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001927476.5 XP_001927511.1 - 659 137
GenBank mRNA AB271955.1 - 2 - 3 511 137
AK396352.1 - 2 - 3 598 137

View Full Product Details